Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSPA14
Entrez Gene ID
51182
Gene Name
heat shock protein family A (Hsp70) member 14
Gene Aliases
HSP70-4, HSP70L1
Chromosome Location
Chr.10: 14838160 - 14871741 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016299.3 NP_057383.2 9 - 10 1128 126
GenBank mRNA AF143723.1 - 9 - 10 937 126
AK225265.1 - 9 - 10 972 126
AK225651.1 - 9 - 10 972 126
AK226065.1 - 2 - 3 3801 126
AK292323.1 - 9 - 10 936 126
BC119690.1 - 9 - 10 947 126

View Full Product Details