Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AHNAK
Entrez Gene ID
79026
Gene Name
AHNAK nucleoprotein
Gene Aliases
AHNAKRS
Chromosome Location
Chr.11: 62433542 - 62546860 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001620.2 NP_001611.1 1 - 2 199 71
NM_024060.3 NP_076965.2 1 - 2 199 71
XM_005274245.1 XP_005274302.1 1 - 2 141 71
XM_017018270.1 XP_016873759.1 1 - 2 141 71
XM_017018271.1 XP_016873760.1 1 - 2 141 71
XM_017018272.1 XP_016873761.1 1 - 2 141 71
XM_017018273.1 XP_016873762.1 1 - 2 141 71
GenBank mRNA AK300376.1 - 1 - 2 174 71
BC000926.1 - 1 - 2 138 71
BC012477.1 - 1 - 2 135 71
BC128460.1 - 1 - 2 188 71
CB129722.1 - 1 - 2 199 71

View Full Product Details