The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033109.4 | NP_149100.2 | 11 - 12 | 1062 | 129 |
XM_005264629.2 | XP_005264686.1 | 11 - 12 | 992 | 129 | |
XM_017005172.1 | XP_016860661.1 | 10 - 11 | 942 | 129 | |
GenBank mRNA | AJ458465.1 | - | 11 - 12 | 993 | 129 |
AK307589.1 | - | - | 992 | 129 | |
AY027528.1 | - | 11 - 12 | 989 | 129 | |
BC053660.1 | - | 11 - 12 | 976 | 129 |