The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033109.4 | NP_149100.2 | 24 - 25 | 2101 | 86 |
XM_005264629.2 | XP_005264686.1 | 24 - 25 | 2031 | 86 | |
XM_017005172.1 | XP_016860661.1 | 23 - 24 | 1981 | 86 | |
GenBank mRNA | AJ458465.1 | - | 24 - 25 | 2032 | 86 |
AK307589.1 | - | - | 2031 | 86 | |
AY290863.1 | - | 6 - 7 | 491 | 86 | |
BC000862.2 | - | 6 - 7 | 465 | 86 | |
BC005986.1 | - | 6 - 7 | 454 | 86 | |
BC053660.1 | - | 24 - 25 | 2015 | 86 | |
CR749867.1 | - | 5 - 6 | 404 | 86 |