The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080975.1 | NP_001074444.1 | 8 - 9 | 1283 | 72 |
NM_004726.2 | NP_004717.2 | 8 - 9 | 1286 | 72 | |
XM_005274625.3 | XP_005274682.1 | 8 - 9 | 1250 | 72 | |
XM_005274626.3 | XP_005274683.1 | 8 - 9 | 1247 | 72 | |
XM_011545603.2 | XP_011543905.1 | 8 - 9 | 1250 | 72 | |
XM_011545604.2 | XP_011543906.1 | 8 - 9 | 1247 | 72 | |
XM_011545605.2 | XP_011543907.1 | 8 - 9 | 1250 | 72 | |
XM_011545606.2 | XP_011543908.1 | 8 - 9 | 1250 | 72 | |
XM_011545607.2 | XP_011543909.1 | 6 - 7 | 974 | 72 | |
XM_011545609.2 | XP_011543911.1 | 8 - 9 | 898 | 72 | |
XM_017029955.1 | XP_016885444.1 | 8 - 9 | 1247 | 72 | |
XM_017029956.1 | XP_016885445.1 | 8 - 9 | 1250 | 72 | |
XM_017029957.1 | XP_016885446.1 | 8 - 9 | 1250 | 72 | |
XM_017029958.1 | XP_016885447.1 | 8 - 9 | 1250 | 72 | |
GenBank mRNA | AB208937.1 | - | 7 - 8 | 838 | 72 |
AF010233.1 | - | 8 - 9 | 1286 | 72 | |
AF511533.1 | - | 8 - 9 | 1112 | 72 | |
AF512951.1 | - | 8 - 9 | 1112 | 72 | |
AK298913.1 | - | 8 - 9 | 897 | 72 |