The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321439.1 | NP_001308368.1 | 2 - 3 | 213 | 82 |
NM_001321440.1 | NP_001308369.1 | 2 - 3 | 213 | 82 | |
NM_024029.4 | NP_076934.1 | 2 - 3 | 484 | 82 | |
XM_011528270.2 | XP_011526572.1 | 2 - 3 | 213 | 82 | |
XM_011528271.2 | XP_011526573.1 | 2 - 3 | 161 | 82 | |
GenBank mRNA | BC000056.2 | - | 2 - 3 | 132 | 82 |
BC013014.2 | - | 2 - 3 | 153 | 82 | |
BM554546.1 | - | 2 - 3 | 131 | 82 | |
HY020827.1 | - | 2 - 3 | 218 | 82 |