The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321439.1 | NP_001308368.1 | 6 - 7 | 677 | 78 |
NM_024029.4 | NP_076934.1 | 6 - 7 | 948 | 78 | |
XM_011528270.2 | XP_011526572.1 | 6 - 7 | 677 | 78 | |
XM_011528271.2 | XP_011526573.1 | 5 - 6 | 538 | 78 | |
GenBank mRNA | BC000056.2 | - | 6 - 7 | 596 | 78 |
BC013014.2 | - | 6 - 7 | 617 | 78 |