The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321439.1 | NP_001308368.1 | 8 - 9 | 1016 | 93 |
NM_001321440.1 | NP_001308369.1 | 7 - 8 | 899 | 93 | |
NM_024029.4 | NP_076934.1 | 8 - 9 | 1287 | 93 | |
XM_011528270.2 | XP_011526572.1 | 8 - 9 | 1016 | 93 | |
XM_011528271.2 | XP_011526573.1 | 7 - 8 | 877 | 93 | |
GenBank mRNA | BC000056.2 | - | 8 - 9 | 935 | 93 |
BC013014.2 | - | 8 - 9 | 956 | 93 | |
BE275116.1 | - | 3 - 4 | 379 | 93 |