Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCA10
Entrez Gene ID
10349
Gene Name
ATP binding cassette subfamily A member 10
Gene Aliases
EST698739
Chromosome Location
Chr.17: 69148007 - 69244815 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080282.3 NP_525021.3 21 - 22 3367 69
GenBank mRNA AL832004.2 - 21 - 22 3073 69
AY028900.1 - 21 - 22 3367 69
AY247065.1 - 21 - 22 3398 69
BC144124.1 - 18 - 19 2641 69

View Full Product Details