Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCA10
Entrez Gene ID
10349
Gene Name
ATP binding cassette subfamily A member 10
Gene Aliases
EST698739
Chromosome Location
Chr.17: 69148007 - 69244815 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080282.3 NP_525021.3 25 - 26 3927 105
GenBank mRNA AL832004.2 - 26 - 27 3722 105
AY028900.1 - 25 - 26 3927 105
AY247065.1 - 25 - 26 3958 105
BC136460.1 - 22 - 23 3132 105
BC144124.1 - 22 - 23 3201 105

View Full Product Details