The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080282.3 | NP_525021.3 | 4 - 5 | 914 | 151 |
GenBank mRNA | AK310858.1 | - | 3 - 4 | 444 | 151 |
AL832004.2 | - | 3 - 4 | 422 | 151 | |
AY028900.1 | - | 4 - 5 | 914 | 151 | |
AY247065.1 | - | 4 - 5 | 945 | 151 | |
BC136460.1 | - | 1 - 2 | 119 | 151 | |
BC144124.1 | - | 1 - 2 | 119 | 151 |