The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006267.4 | NP_006258.3 | 20 - 21 | 7973 | 146 |
XM_005264004.2 | XP_005264061.1 | 20 - 21 | 7936 | 146 | |
GenBank mRNA | AB209483.1 | - | 17 - 18 | 7589 | 146 |
AK025711.1 | - | 1 - 2 | 2149 | 146 | |
D42063.1 | - | 20 - 21 | 7974 | 146 |