The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006267.4 | NP_006258.3 | 26 - 27 | 8890 | 155 |
XM_005264002.2 | XP_005264059.1 | 27 - 28 | 9027 | 155 | |
XM_005264003.2 | XP_005264060.1 | 27 - 28 | 8931 | 155 | |
XM_005264004.2 | XP_005264061.1 | 26 - 27 | 8877 | 155 | |
XM_005264005.4 | XP_005264062.1 | 27 - 28 | 9015 | 155 | |
XM_005264007.2 | XP_005264064.1 | 27 - 28 | 6027 | 155 | |
XM_011511575.2 | XP_011509877.1 | 27 - 28 | 8930 | 155 | |
XM_011511576.2 | XP_011509878.1 | 28 - 29 | 8778 | 155 | |
XM_011511578.2 | XP_011509880.1 | 27 - 28 | 6069 | 155 | |
GenBank mRNA | AB209483.1 | - | 23 - 24 | 8506 | 155 |
AK025711.1 | - | 7 - 8 | 3066 | 155 | |
AY170822.1 | - | 7 - 8 | 993 | 155 | |
D42063.1 | - | 26 - 27 | 8891 | 155 |