The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145296.1 | NP_660339.1 | 4 - 5 | 549 | 60 |
XM_005258620.2 | XP_005258677.1 | 4 - 5 | 531 | 60 | |
XM_017026452.1 | XP_016881941.1 | 4 - 5 | 458 | 60 | |
GenBank mRNA | AF363368.1 | - | 4 - 5 | 549 | 60 |
AK313028.1 | - | 4 - 5 | 531 | 60 | |
BC113963.1 | - | 1 - 2 | 122 | 60 |