Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM33
Entrez Gene ID
51592
Gene Name
tripartite motif containing 33
Gene Aliases
ECTO, PTC7, RFG7, TF1G, TIF1G, TIF1GAMMA, TIFGAMMA
Chromosome Location
Chr.1: 114392777 - 114511160 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015906.3 NP_056990.3 10 - 11 1949 76
NM_033020.2 NP_148980.2 10 - 11 1949 76
XM_005270936.3 XP_005270993.1 10 - 11 1870 76
XM_005270937.3 XP_005270994.1 10 - 11 1870 76
XM_011541568.2 XP_011539870.1 10 - 11 1870 76
XM_017001452.1 XP_016856941.1 8 - 9 1224 76
XM_017001453.1 XP_016856942.1 8 - 9 1224 76
GenBank mRNA AB029036.1 - 10 - 11 1878 76
AF119043.1 - 10 - 11 1949 76
AF220136.1 - 10 - 11 1949 76
AF220137.1 - 10 - 11 1949 76
AJ132948.1 - 10 - 11 1640 76
AK023434.1 - 7 - 8 1090 76
AK301997.1 - 4 - 5 555 76

View Full Product Details