The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015906.3 | NP_056990.3 | 6 - 7 | 120 | |
NM_033020.2 | NP_148980.2 | 6 - 7 | 120 | ||
XM_005270936.3 | XP_005270993.1 | 6 - 7 | 120 | ||
XM_005270937.3 | XP_005270994.1 | 6 - 7 | 120 | ||
XM_011541568.2 | XP_011539870.1 | 6 - 7 | 120 | ||
XM_017001454.1 | XP_016856943.1 | 6 - 7 | 120 | ||
GenBank mRNA | AB029036.1 | - | 6 - 7 | 1166 | 120 |
AF119043.1 | - | 6 - 7 | 120 | ||
AF220136.1 | - | 6 - 7 | 120 | ||
AF220137.1 | - | 6 - 7 | 120 | ||
AJ132948.1 | - | 6 - 7 | 928 | 120 | |
AK023434.1 | - | 3 - 4 | 120 |