The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000552.4 | NP_000543.2 | 23 - 24 | 3365 | 131 |
GenBank mRNA | M10321.1 | - | 7 - 8 | 894 | 131 |
X04146.1 | - | 23 - 24 | 3339 | 131 | |
X04385.1 | - | 23 - 24 | 3209 | 131 |