The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199799.1 | NP_001186728.1 | 1 - 2 | 259 | 86 |
NM_001199800.1 | NP_001186729.1 | 1 - 2 | 259 | 86 | |
NM_175924.3 | NP_787120.1 | 1 - 2 | 259 | 86 | |
XM_011512738.2 | XP_011511040.1 | 1 - 2 | 239 | 86 | |
GenBank mRNA | AK129974.1 | - | 1 - 2 | 259 | 86 |
AY134857.1 | - | 1 - 2 | 177 | 86 | |
AY672837.1 | - | 1 - 2 | 101 | 86 | |
AY672839.1 | - | 1 - 2 | 101 | 86 | |
BC044240.1 | - | 1 - 2 | 162 | 86 |