Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBA6
Entrez Gene ID
55236
Gene Name
ubiquitin like modifier activating enzyme 6
Gene Aliases
E1-L2, MOP-4, UBE1L2
Chromosome Location
Chr.4: 67615761 - 67701171 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018227.5 NP_060697.4 32 - 33 3066 103
GenBank mRNA AB014773.1 - 32 - 33 3014 103
AK001670.1 - 14 - 15 1415 103
AK027020.1 - 2 - 3 190 103
AK094164.1 - 24 - 25 2272 103
AK314371.1 - 32 - 33 3043 103
AL832015.2 - 32 - 33 3035 103
AL832458.2 - 32 - 33 3035 103
AY359880.1 - 19 - 20 2387 103
BC126484.1 - 32 - 33 3061 103
BC126486.1 - 32 - 33 3061 103
EF623993.1 - 32 - 33 3014 103

View Full Product Details