The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005744.3 | NP_005735.2 | 12 - 13 | 1783 | 100 |
XM_011521438.2 | XP_011519740.1 | 11 - 12 | 1759 | 100 | |
XM_011521439.2 | XP_011519741.1 | 13 - 14 | 1297 | 100 | |
GenBank mRNA | AB014774.1 | - | 3 - 4 | 343 | 100 |
AB209432.1 | - | 13 - 14 | 1412 | 100 | |
AF072832.1 | - | 12 - 13 | 1469 | 100 | |
AJ009771.1 | - | 5 - 6 | 579 | 100 | |
AJ130976.1 | - | 12 - 13 | 1187 | 100 | |
AJ243190.1 | - | 12 - 13 | 1783 | 100 | |
AK312715.1 | - | 12 - 13 | 1782 | 100 | |
BC051877.1 | - | 12 - 13 | 1718 | 100 |