Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCT
Entrez Gene ID
6343
Gene Name
secretin
Gene Aliases
-
Chromosome Location
Chr.11: 626095 - 627692 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021920.3 NP_068739.1 3 - 4 365 96
XM_017018109.1 XP_016873598.1 3 - 4 361 96
GenBank mRNA HY019819.1 - 3 - 4 365 96
HY243621.1 - 1 - 2 96

View Full Product Details