The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190728.1 | NP_001177657.1 | 16 - 17 | 1562 | 86 |
NM_001190729.1 | NP_001177658.1 | 17 - 18 | 1825 | 86 | |
NM_001322320.1 | NP_001309249.1 | 17 - 18 | 1788 | 86 | |
NM_001322323.1 | NP_001309252.1 | 16 - 17 | 1735 | 86 | |
NM_001322325.1 | NP_001309254.1 | 19 - 20 | 2035 | 86 | |
XM_006717328.3 | XP_006717391.2 | 20 - 21 | 2009 | 86 | |
XM_011519225.1 | XP_011517527.1 | 19 - 20 | 1955 | 86 | |
XM_011519226.1 | XP_011517528.1 | 19 - 20 | 1934 | 86 | |
XM_011519227.1 | XP_011517529.1 | 18 - 19 | 1907 | 86 | |
XM_011519229.1 | XP_011517531.1 | 20 - 21 | 2015 | 86 | |
XM_011519230.2 | XP_011517532.1 | 20 - 21 | 2009 | 86 | |
XM_011519231.1 | XP_011517533.1 | 17 - 18 | 1748 | 86 | |
XM_011519232.1 | XP_011517534.1 | 17 - 18 | 1745 | 86 | |
XM_011519234.2 | XP_011517536.1 | 19 - 20 | 2092 | 86 | |
XM_011519235.2 | XP_011517537.1 | 20 - 21 | 2183 | 86 | |
XM_017015343.1 | XP_016870832.1 | 20 - 21 | 2002 | 86 | |
XM_017015344.1 | XP_016870833.1 | 17 - 18 | 1775 | 86 | |
XM_017015349.1 | XP_016870838.1 | 16 - 17 | 1643 | 86 | |
GenBank mRNA | AF221098.1 | - | 16 - 17 | 1562 | 86 |
AK299149.1 | - | 17 - 18 | 1766 | 86 |