The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276718.1 | NP_001263647.1 | 1 - 2 | 709 | 64 |
GenBank mRNA | AK131522.1 | - | 1 - 2 | 709 | 64 |