Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CCDC33
- Entrez Gene ID
-
80125
- Gene Name
- coiled-coil domain containing 33
- Gene Aliases
-
CT61, HP11097
- Chromosome Location
-
Chr.15: 74202705 - 74336472 on Build GRCh38
View Full Product Details