The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285460.1 | NP_001272389.1 | 13 - 14 | 1515 | 180 |
NM_001285461.1 | NP_001272390.1 | 3 - 4 | 346 | 180 | |
NM_001285462.1 | NP_001272391.1 | 3 - 4 | 397 | 180 | |
NM_006633.3 | NP_006624.2 | 14 - 15 | 1909 | 180 | |
XM_005248410.2 | XP_005248467.1 | 14 - 15 | 1759 | 180 | |
XM_005248414.1 | XP_005248471.1 | 3 - 4 | 354 | 180 | |
XM_011543108.1 | XP_011541410.1 | 3 - 4 | 346 | 180 | |
XM_017008960.1 | XP_016864449.1 | 14 - 15 | 1858 | 180 | |
GenBank mRNA | AB208856.1 | - | 3 - 4 | 358 | 180 |
AK291066.1 | - | 14 - 15 | 1909 | 180 | |
AK302510.1 | - | 13 - 14 | 1515 | 180 | |
AK302599.1 | - | 3 - 4 | 397 | 180 | |
AK302981.1 | - | 2 - 3 | 132 | 180 | |
AK303054.1 | - | 3 - 4 | 366 | 180 | |
AK307555.1 | - | 14 - 15 | 1683 | 180 | |
AK311619.1 | - | 3 - 4 | 363 | 180 | |
AY351902.1 | - | 3 - 4 | 322 | 180 | |
DB071610.1 | - | 3 - 4 | 346 | 180 | |
DB075371.1 | - | 3 - 4 | 336 | 180 | |
U51903.1 | - | 14 - 15 | 1840 | 180 |