The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285460.1 | NP_001272389.1 | 28 - 29 | 3658 | 70 |
NM_001285461.1 | NP_001272390.1 | 17 - 18 | 2318 | 70 | |
NM_001285462.1 | NP_001272391.1 | 17 - 18 | 2369 | 70 | |
NM_006633.3 | NP_006624.2 | 29 - 30 | 4052 | 70 | |
XM_005248410.2 | XP_005248467.1 | 29 - 30 | 3902 | 70 | |
XM_005248414.1 | XP_005248471.1 | 18 - 19 | 2497 | 70 | |
XM_011543108.1 | XP_011541410.1 | 18 - 19 | 2489 | 70 | |
XM_017008960.1 | XP_016864449.1 | 28 - 29 | 3830 | 70 | |
GenBank mRNA | AB208856.1 | - | 18 - 19 | 2501 | 70 |
AK291066.1 | - | 29 - 30 | 4052 | 70 | |
AK302510.1 | - | 28 - 29 | 3658 | 70 | |
AK302981.1 | - | 16 - 17 | 2104 | 70 | |
AK303054.1 | - | 17 - 18 | 2338 | 70 | |
AK307626.1 | - | 2 - 3 | 289 | 70 | |
AY351902.1 | - | 17 - 18 | 2294 | 70 | |
U51903.1 | - | 29 - 30 | 3983 | 70 |