The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172070.3 | NP_742067.3 | 37 - 38 | 5410 | 116 |
XM_005246299.4 | XP_005246356.1 | 38 - 39 | 5500 | 116 | |
XM_005246300.4 | XP_005246357.1 | 38 - 39 | 5443 | 116 | |
XM_006712268.3 | XP_006712331.1 | 38 - 39 | 5497 | 116 | |
XM_006712269.3 | XP_006712332.1 | 37 - 38 | 5422 | 116 | |
XM_006712270.3 | XP_006712333.2 | 37 - 38 | 5356 | 116 | |
XM_006712271.3 | XP_006712334.1 | 37 - 38 | 5463 | 116 | |
GenBank mRNA | AB095944.1 | - | 8 - 9 | 1520 | 116 |
AK094741.1 | - | 1 - 2 | 182 | 116 | |
AK126998.1 | - | 15 - 16 | 2139 | 116 | |
AK127553.1 | - | 11 - 12 | 1416 | 116 | |
AK128490.1 | - | 21 - 22 | 2890 | 116 | |
AK303062.1 | - | 12 - 13 | 1510 | 116 | |
AL834144.1 | - | 9 - 10 | 1117 | 116 | |
BX649036.1 | - | 13 - 14 | 1646 | 116 | |
CR749469.1 | - | 15 - 16 | 1912 | 116 |