Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRRG2
Entrez Gene ID
5639
Gene Name
proline rich and Gla domain 2
Gene Aliases
PRGP2
Chromosome Location
Chr.19: 49580617 - 49591008 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000951.2 NP_000942.1 2 - 3 242 94
XM_006723286.2 XP_006723349.1 3 - 4 245 94
GenBank mRNA AF009243.1 - 1 - 2 86 94
CR456846.1 - 1 - 2 77 94

View Full Product Details