The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032233.2 | NP_115609.2 | 10 - 11 | 1265 | 73 |
XM_005268127.3 | XP_005268184.1 | 9 - 10 | 1118 | 73 | |
XM_011537231.2 | XP_011535533.2 | 10 - 11 | 1217 | 73 | |
XM_011537232.2 | XP_011535534.1 | 10 - 11 | 2395 | 73 | |
XM_011537233.2 | XP_011535535.1 | 8 - 9 | 1043 | 73 | |
XM_011537234.1 | XP_011535536.1 | 9 - 10 | 985 | 73 | |
XM_017021699.1 | XP_016877188.1 | 9 - 10 | 1113 | 73 | |
XM_017021700.1 | XP_016877189.1 | 10 - 11 | 1301 | 73 | |
XM_017021701.1 | XP_016877190.1 | 8 - 9 | 1093 | 73 | |
XM_017021702.1 | XP_016877191.1 | 7 - 8 | 912 | 73 | |
GenBank mRNA | AK026680.1 | - | 9 - 10 | 1019 | 73 |
AK302882.1 | - | 10 - 11 | 2608 | 73 | |
AL359581.2 | - | 5 - 6 | 455 | 73 | |
BC142995.1 | - | 10 - 11 | 1277 | 73 | |
BX161471.1 | - | 10 - 11 | 1255 | 73 |