The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032233.2 | NP_115609.2 | 5 - 6 | 590 | 87 |
NM_199123.1 | NP_954574.1 | 5 - 6 | 590 | 87 | |
XM_005268127.3 | XP_005268184.1 | 4 - 5 | 443 | 87 | |
XM_011537231.2 | XP_011535533.2 | 5 - 6 | 542 | 87 | |
XM_011537232.2 | XP_011535534.1 | 5 - 6 | 1720 | 87 | |
XM_011537233.2 | XP_011535535.1 | 5 - 6 | 542 | 87 | |
XM_011537234.1 | XP_011535536.1 | 4 - 5 | 310 | 87 | |
XM_011537235.1 | XP_011535537.1 | 5 - 6 | 542 | 87 | |
XM_017021699.1 | XP_016877188.1 | 4 - 5 | 438 | 87 | |
XM_017021700.1 | XP_016877189.1 | 5 - 6 | 626 | 87 | |
XM_017021701.1 | XP_016877190.1 | 5 - 6 | 592 | 87 | |
XM_017021702.1 | XP_016877191.1 | 4 - 5 | 411 | 87 | |
GenBank mRNA | AK026680.1 | - | 4 - 5 | 344 | 87 |
BC009054.1 | - | 2 - 3 | 216 | 87 | |
BC066931.1 | - | 5 - 6 | 491 | 87 | |
BC120967.1 | - | 5 - 6 | 445 | 87 | |
BC120968.2 | - | 5 - 6 | 445 | 87 | |
BC127624.1 | - | 5 - 6 | 445 | 87 | |
BC127625.1 | - | 5 - 6 | 445 | 87 | |
BC142995.1 | - | 5 - 6 | 602 | 87 | |
BC148251.1 | - | 4 - 5 | 441 | 87 | |
BX161441.1 | - | 5 - 6 | 489 | 87 | |
BX161471.1 | - | 5 - 6 | 580 | 87 | |
CB988965.1 | - | 5 - 6 | 617 | 87 |