The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032233.2 | NP_115609.2 | 7 - 8 | 899 | 79 |
NM_199123.1 | NP_954574.1 | 7 - 8 | 899 | 79 | |
XM_005268127.3 | XP_005268184.1 | 6 - 7 | 752 | 79 | |
XM_011537231.2 | XP_011535533.2 | 7 - 8 | 851 | 79 | |
XM_011537232.2 | XP_011535534.1 | 7 - 8 | 2029 | 79 | |
XM_011537234.1 | XP_011535536.1 | 6 - 7 | 619 | 79 | |
XM_011537235.1 | XP_011535537.1 | 7 - 8 | 851 | 79 | |
XM_017021699.1 | XP_016877188.1 | 6 - 7 | 747 | 79 | |
XM_017021700.1 | XP_016877189.1 | 7 - 8 | 935 | 79 | |
GenBank mRNA | AK026680.1 | - | 6 - 7 | 653 | 79 |
AK302882.1 | - | 7 - 8 | 2242 | 79 | |
AL359581.2 | - | 2 - 3 | 89 | 79 | |
BC009054.1 | - | 4 - 5 | 525 | 79 | |
BC066931.1 | - | 7 - 8 | 800 | 79 | |
BC120967.1 | - | 7 - 8 | 754 | 79 | |
BC120968.2 | - | 7 - 8 | 754 | 79 | |
BC127624.1 | - | 7 - 8 | 754 | 79 | |
BC127625.1 | - | 7 - 8 | 754 | 79 | |
BC142995.1 | - | 7 - 8 | 911 | 79 | |
BC148251.1 | - | 6 - 7 | 750 | 79 | |
BX161441.1 | - | 7 - 8 | 798 | 79 | |
BX161471.1 | - | 7 - 8 | 889 | 79 |