Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTSL3
Entrez Gene ID
57188
Gene Name
ADAMTS like 3
Gene Aliases
ADAMTSL-3
Chromosome Location
Chr.15: 83654086 - 84039842 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301110.1 NP_001288039.1 19 - 20 2715 73
NM_207517.2 NP_997400.2 19 - 20 2715 73
XM_011521822.2 XP_011520124.1 20 - 21 2787 73
XM_011521823.2 XP_011520125.1 20 - 21 2787 73
XM_011521824.2 XP_011520126.1 20 - 21 2787 73
XM_011521825.2 XP_011520127.1 20 - 21 2787 73
XM_017022434.1 XP_016877923.1 20 - 21 2787 73
XM_017022435.1 XP_016877924.1 19 - 20 2583 73
GenBank mRNA AB033059.1 - 3 - 4 489 73
AK308845.1 - 19 - 20 2706 73
BC128389.1 - 19 - 20 2713 73
BC128390.1 - 19 - 20 2713 73

View Full Product Details