Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NIPBL
Entrez Gene ID
25836
Gene Name
NIPBL, cohesin loading factor
Gene Aliases
CDLS, CDLS1, IDN3, IDN3-B, Scc2
Chromosome Location
Chr.5: 36876759 - 37065819 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015384.4 NP_056199.2 26 - 27 5720 112
NM_133433.3 NP_597677.2 26 - 27 5720 112
XM_005248280.3 XP_005248337.1 26 - 27 5709 112
XM_005248282.4 XP_005248339.3 20 - 21 5211 112
XM_006714467.2 XP_006714530.1 26 - 27 5709 112
XM_006714468.2 XP_006714531.1 25 - 26 5511 112
XM_011514015.1 XP_011512317.1 26 - 27 5709 112
XM_017009329.1 XP_016864818.1 26 - 27 5709 112
XM_017009330.1 XP_016864819.1 18 - 19 7983 112
XM_017009331.1 XP_016864820.1 25 - 26 4083 112
GenBank mRNA AJ627032.1 - 26 - 27 5689 112
AJ640137.1 - 26 - 27 5689 112
KJ807791.1 - 12 - 13 1507 112

View Full Product Details