The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015384.4 | NP_056199.2 | 7 - 8 | 1265 | 79 |
NM_133433.3 | NP_597677.2 | 7 - 8 | 1265 | 79 | |
XM_005248280.3 | XP_005248337.1 | 7 - 8 | 1254 | 79 | |
XM_006714467.2 | XP_006714530.1 | 7 - 8 | 1254 | 79 | |
XM_006714468.2 | XP_006714531.1 | 7 - 8 | 1254 | 79 | |
XM_011514015.1 | XP_011512317.1 | 7 - 8 | 1254 | 79 | |
XM_017009329.1 | XP_016864818.1 | 7 - 8 | 1254 | 79 | |
XM_017009331.1 | XP_016864820.1 | 7 - 8 | 1254 | 79 | |
GenBank mRNA | AJ627032.1 | - | 7 - 8 | 1234 | 79 |
AJ640137.1 | - | 7 - 8 | 1234 | 79 | |
AK126804.1 | - | 3 - 4 | 386 | 79 | |
AK290347.1 | - | 7 - 8 | 908 | 79 | |
BC146821.1 | - | 7 - 8 | 1174 | 79 | |
BX538177.1 | - | 7 - 8 | 883 | 79 | |
BX538178.1 | - | 7 - 8 | 892 | 79 | |
BX640644.1 | - | 7 - 8 | 1141 | 79 |