The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178030.1 | NP_001171501.1 | 9 - 10 | 1520 | 55 |
NM_001178031.1 | NP_001171502.1 | 9 - 10 | 1302 | 55 | |
NM_004630.3 | NP_004621.2 | 9 - 10 | 1523 | 55 | |
NM_201995.2 | NP_973724.1 | 9 - 10 | 1523 | 55 | |
NM_201997.2 | NP_973726.2 | 9 - 10 | 1523 | 55 | |
NM_201998.2 | NP_973727.1 | 9 - 10 | 1523 | 55 | |
NR_033649.1 | 8 - 9 | 1447 | 55 | ||
NR_033650.1 | 8 - 9 | 1447 | 55 | ||
XM_011545244.1 | XP_011543546.1 | 9 - 10 | 1195 | 55 | |
XM_011545245.1 | XP_011543547.1 | 9 - 10 | 1195 | 55 | |
XM_011545246.1 | XP_011543548.1 | 9 - 10 | 1195 | 55 | |
XM_011545247.1 | XP_011543549.1 | 9 - 10 | 1195 | 55 | |
XM_011545248.1 | XP_011543550.1 | 9 - 10 | 1195 | 55 | |
XM_017018244.1 | XP_016873733.1 | 9 - 10 | 1195 | 55 | |
XM_017018245.1 | XP_016873734.1 | 9 - 10 | 1276 | 55 | |
XM_017018246.1 | XP_016873735.1 | 9 - 10 | 1195 | 55 | |
XM_017018247.1 | XP_016873736.1 | 9 - 10 | 1238 | 55 | |
XM_017018248.1 | XP_016873737.1 | 9 - 10 | 1238 | 55 | |
XM_017018249.1 | XP_016873738.1 | 9 - 10 | 3688 | 55 | |
XM_017018250.1 | XP_016873739.1 | 8 - 9 | 1372 | 55 | |
XM_017018251.1 | XP_016873740.1 | 8 - 9 | 1227 | 55 | |
GenBank mRNA | AK293753.1 | - | 9 - 10 | 1302 | 55 |
AK296237.1 | - | 9 - 10 | 1194 | 55 | |
AK299705.1 | - | 7 - 8 | 933 | 55 | |
AK301803.1 | - | 8 - 9 | 1372 | 55 | |
BC008080.2 | - | 9 - 10 | 1219 | 55 | |
BC008724.2 | - | 9 - 10 | 1219 | 55 | |
BC020217.1 | - | 9 - 10 | 1422 | 55 | |
BC034451.1 | - | 9 - 10 | 1448 | 55 | |
BC038446.1 | - | 9 - 10 | 1543 | 55 | |
D26120.1 | - | 9 - 10 | 1453 | 55 | |
Y08765.1 | - | 9 - 10 | 1113 | 55 | |
Y08766.1 | - | 9 - 10 | 1365 | 55 |