Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPACA1
Entrez Gene ID
81833
Gene Name
sperm acrosome associated 1
Gene Aliases
SAMP32
Chromosome Location
Chr.6: 88046907 - 88066832 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030960.2 NP_112222.1 2 - 3 380 121
GenBank mRNA AF203447.1 - 2 - 3 121
BC029488.1 - 2 - 3 380 121

View Full Product Details