The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130720.2 | NP_001124192.1 | 14 - 15 | 1691 | 75 |
NM_001130722.2 | NP_001124194.2 | 14 - 15 | 1549 | 75 | |
NM_001130723.2 | NP_001124195.1 | 14 - 15 | 1694 | 75 | |
NM_015328.3 | NP_056143.1 | 14 - 15 | 1694 | 75 | |
XM_005250237.1 | XP_005250294.1 | 14 - 15 | 1697 | 75 | |
XM_011515987.2 | XP_011514289.1 | 15 - 16 | 2668 | 75 | |
XM_011515988.2 | XP_011514290.1 | 15 - 16 | 2635 | 75 | |
XM_017011904.1 | XP_016867393.1 | 14 - 15 | 1412 | 75 | |
XM_017011905.1 | XP_016867394.1 | 14 - 15 | 1572 | 75 | |
XM_017011906.1 | XP_016867395.1 | 15 - 16 | 1588 | 75 | |
XM_017011907.1 | XP_016867396.1 | 15 - 16 | 1396 | 75 | |
GenBank mRNA | AB020635.1 | - | 14 - 15 | 1655 | 75 |
AK025372.1 | - | 14 - 15 | 1593 | 75 | |
AK295851.1 | - | 14 - 15 | 1383 | 75 | |
BC008349.1 | - | 14 - 15 | 1633 | 75 | |
BC024325.1 | - | 14 - 15 | 1685 | 75 |