Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AHCYL2
Entrez Gene ID
23382
Gene Name
adenosylhomocysteinase like 2
Gene Aliases
ADOHCYASE3, IRBIT2
Chromosome Location
Chr.7: 129225014 - 129430211 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130720.2 NP_001124192.1 15 - 16 1768 81
NM_001130722.2 NP_001124194.2 15 - 16 1626 81
NM_001130723.2 NP_001124195.1 15 - 16 1771 81
NM_015328.3 NP_056143.1 15 - 16 1771 81
XM_005250237.1 XP_005250294.1 15 - 16 1774 81
XM_011515987.2 XP_011514289.1 16 - 17 2745 81
XM_011515988.2 XP_011514290.1 16 - 17 2712 81
XM_017011904.1 XP_016867393.1 15 - 16 1489 81
XM_017011905.1 XP_016867394.1 15 - 16 1649 81
XM_017011906.1 XP_016867395.1 16 - 17 1665 81
XM_017011907.1 XP_016867396.1 16 - 17 1473 81
GenBank mRNA AB020635.1 - 15 - 16 1732 81
AK025372.1 - 15 - 16 1670 81
AK295851.1 - 15 - 16 1460 81
AK316073.1 - 15 - 16 1626 81
BC008349.1 - 15 - 16 1710 81
BC024325.1 - 15 - 16 1762 81

View Full Product Details