Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO4
Entrez Gene ID
121601
Gene Name
anoctamin 4
Gene Aliases
TMEM16D
Chromosome Location
Chr.12: 100717457 - 101128641 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286615.1 NP_001273544.1 15 - 16 1963 112
NM_001286616.1 NP_001273545.1 14 - 15 1531 112
NM_178826.3 NP_849148.2 14 - 15 1858 112
XM_011537911.2 XP_011536213.1 17 - 18 1958 112
XM_011537912.2 XP_011536214.1 17 - 18 1958 112
XM_011537913.2 XP_011536215.1 16 - 17 1913 112
XM_011537914.2 XP_011536216.1 14 - 15 1469 112
XM_011537915.2 XP_011536217.1 15 - 16 1781 112
XM_011537916.2 XP_011536218.1 15 - 16 1622 112
XM_017018808.1 XP_016874297.1 14 - 15 1469 112
XM_017018809.1 XP_016874298.1 16 - 17 2049 112
XM_017018810.1 XP_016874299.1 14 - 15 1711 112
XM_017018811.1 XP_016874300.1 13 - 14 1666 112
XM_017018812.1 XP_016874301.1 13 - 14 3684 112
XM_017018813.1 XP_016874302.1 12 - 13 1534 112
XM_017018814.1 XP_016874303.1 15 - 16 3923 112
XM_017018815.1 XP_016874304.1 15 - 16 3923 112
XM_017018816.1 XP_016874305.1 15 - 16 2006 112
XM_017018817.1 XP_016874306.1 15 - 16 3958 112
GenBank mRNA AK091540.1 - 5 - 6 479 112
AK091591.1 - 14 - 15 1647 112
AK293810.1 - 15 - 16 1601 112
BC109308.2 - 14 - 15 1516 112

View Full Product Details