The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001265614.2 | NP_001252543.1 | 4 - 5 | 875 | 90 |
XM_005251688.1 | XP_005251745.1 | 4 - 5 | 630 | 90 | |
XM_006716948.2 | XP_006717011.1 | 4 - 5 | 1031 | 90 | |
XM_006716949.3 | XP_006717012.1 | 4 - 5 | 891 | 90 | |
GenBank mRNA | AB067483.1 | - | 4 - 5 | 874 | 90 |