Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HMCN2
Entrez Gene ID
256158
Gene Name
hemicentin 2
Gene Aliases
-
Chromosome Location
Chr.9: 130265742 - 130434123 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291815.1 NP_001278744.1 10 - 11 1539 112
XM_011518465.2 XP_011516767.1 10 - 11 1676 112
XM_011518466.2 XP_011516768.1 10 - 11 1676 112
XM_011518467.2 XP_011516769.1 10 - 11 1676 112
XM_011518468.2 XP_011516770.1 10 - 11 1676 112
XM_011518469.2 XP_011516771.1 10 - 11 1676 112
XM_011518470.2 XP_011516772.1 10 - 11 1676 112

View Full Product Details