The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025055.4 | NP_079331.3 | 4 - 5 | 860 | 63 |
XM_005254692.1 | XP_005254749.1 | 4 - 5 | 860 | 63 | |
XM_011522085.2 | XP_011520387.1 | 5 - 6 | 1705 | 63 | |
XM_011522086.1 | XP_011520388.1 | 4 - 5 | 553 | 63 | |
XM_011522087.2 | XP_011520389.1 | 5 - 6 | 691 | 63 | |
XM_011522088.1 | XP_011520390.1 | 4 - 5 | 561 | 63 | |
XM_017022623.1 | XP_016878112.1 | 5 - 6 | 1706 | 63 | |
XM_017022624.1 | XP_016878113.1 | 5 - 6 | 1705 | 63 | |
XM_017022625.1 | XP_016878114.1 | 5 - 6 | 1705 | 63 | |
XM_017022626.1 | XP_016878115.1 | 5 - 6 | 1705 | 63 | |
XM_017022627.1 | XP_016878116.1 | 5 - 6 | 1705 | 63 | |
XM_017022628.1 | XP_016878117.1 | 5 - 6 | 1705 | 63 | |
XM_017022629.1 | XP_016878118.1 | 7 - 8 | 1446 | 63 | |
XM_017022630.1 | XP_016878119.1 | 5 - 6 | 1707 | 63 | |
XM_017022631.1 | XP_016878120.1 | 5 - 6 | 1706 | 63 | |
GenBank mRNA | AK057417.1 | - | 4 - 5 | 746 | 63 |
BC031684.1 | - | 1 - 2 | 70 | 63 |