Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRIP3
Entrez Gene ID
401262
Gene Name
cysteine rich protein 3
Gene Aliases
TLP, TLP-A, bA480N24.2
Chromosome Location
Chr.6: 43305473 - 43310705 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_206922.2 NP_996805.2 6 - 7 498 88
XM_005249103.3 XP_005249160.1 6 - 7 2411 88
XM_017010856.1 XP_016866345.1 5 - 6 2279 88
XM_017010857.1 XP_016866346.1 4 - 5 2207 88
GenBank mRNA AY555741.1 - 6 - 7 498 88
AY555742.1 - 6 - 7 498 88
AY555743.1 - 6 - 7 498 88
CN282994.1 - 5 - 6 445 88

View Full Product Details