The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052900.2 | NP_443132.3 | 7 - 8 | 1267 | 73 |
NM_198123.1 | NP_937756.1 | 8 - 9 | 1579 | 73 | |
NM_198124.1 | NP_937757.1 | 9 - 10 | 1471 | 73 | |
XM_011516816.2 | XP_011515118.1 | 7 - 8 | 1753 | 73 | |
XM_017013009.1 | XP_016868498.1 | 8 - 9 | 2065 | 73 | |
XM_017013012.1 | XP_016868501.1 | 6 - 7 | 1038 | 73 | |
GenBank mRNA | AB114604.1 | - | 8 - 9 | 1579 | 73 |
AB114605.1 | - | 9 - 10 | 1471 | 73 | |
AY210419.1 | - | 7 - 8 | 1138 | 73 |