Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HMGB3
Entrez Gene ID
3149
Gene Name
high mobility group box 3
Gene Aliases
HMG-2a, HMG-4, HMG2A, HMG4
Chromosome Location
Chr.X: 150980508 - 150990775 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301231.1 NP_001288160.1 1 - 2 135 76
GenBank mRNA BF979310.1 - 1 - 2 141 76

View Full Product Details