Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGAP11
Entrez Gene ID
119385
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 11
Gene Aliases
-
Chromosome Location
Chr.10: 86970741 - 87010203 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133447.1 NP_597704.1 8 - 9 1115 106
GenBank mRNA AK097165.1 - 8 - 9 1115 106
BC140762.1 - 4 - 5 361 106

View Full Product Details