The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_026660.1 | 1 - 1 | 241 | 75 | |
GenBank mRNA | AW373666.1 | - | 1 - 1 | 243 | 75 |
CD709306.1 | - | 1 - 1 | 244 | 75 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | armadillo repeat containing 10 | ARMC10 | PNAS-112, PNAS112, PSEC0198, SVH | -42.1kb | Anti-sense | Chr. - 103074727 - 103099763 on Build GRCh38 |
Downstream | DPY19L2 pseudogene 2 | DPY19L2P2 | 32.6kb | Sense | Chr. - 103175013 - 103280466 on Build GRCh38 |