Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC22A25
Entrez Gene ID
387601
Gene Name
solute carrier family 22 member 25
Gene Aliases
HIMTP, UST6
Chromosome Location
Chr.11: 63162264 - 63243523 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199352.3 NP_955384.3 8 - 9 1394 95
XM_017017684.1 XP_016873173.1 12 - 13 2563 95
XM_017017685.1 XP_016873174.1 13 - 14 2691 95
XM_017017686.1 XP_016873175.1 12 - 13 2999 95
XM_017017687.1 XP_016873176.1 11 - 12 3014 95
XM_017017688.1 XP_016873177.1 8 - 9 1952 95
XM_017017689.1 XP_016873178.1 9 - 10 1864 95
XM_017017690.1 XP_016873179.1 11 - 12 1767 95
XM_017017691.1 XP_016873180.1 9 - 10 1220 95
XM_017017692.1 XP_016873181.1 8 - 9 1391 95
XM_017017693.1 XP_016873182.1 7 - 8 982 95
XM_017017694.1 XP_016873183.1 7 - 8 1821 95
XM_017017696.1 XP_016873185.1 9 - 10 1523 95
GenBank mRNA AY437532.1 - 8 - 9 1394 95
BC101315.1 - 9 - 10 1541 95
BC101316.1 - 8 - 9 1394 95
BC101317.1 - 6 - 7 1118 95

View Full Product Details