The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267843.1 | NP_001254772.1 | 6 - 7 | 1028 | 118 |
NM_004898.3 | NP_004889.1 | 5 - 6 | 519 | 118 | |
XM_005265787.2 | XP_005265844.1 | 5 - 6 | 565 | 118 | |
XM_011534409.2 | XP_011532711.1 | 5 - 6 | 974 | 118 | |
XM_011534410.2 | XP_011532712.1 | 4 - 5 | 356 | 118 | |
XM_011534411.2 | XP_011532713.1 | 4 - 5 | 315 | 118 | |
XM_017008854.1 | XP_016864343.1 | 6 - 7 | 905 | 118 | |
XM_017008855.1 | XP_016864344.1 | 2 - 3 | 114 | 118 | |
GenBank mRNA | AB002332.1 | - | 4 - 5 | 353 | 118 |
AB005535.1 | - | 5 - 6 | 723 | 118 | |
AF011568.1 | - | 5 - 6 | 459 | 118 | |
AK223549.1 | - | 5 - 6 | 459 | 118 | |
AK291708.1 | - | 5 - 6 | 458 | 118 | |
BC041878.1 | - | 6 - 7 | 1028 | 118 | |
BC126157.1 | - | 5 - 6 | 435 | 118 | |
BC126159.1 | - | 5 - 6 | 435 | 118 |