Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIN52
Entrez Gene ID
91750
Gene Name
lin-52 DREAM MuvB core complex component
Gene Aliases
C14orf46, c14_5549
Chromosome Location
Chr.14: 74084831 - 74201255 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024674.2 NP_001019845.1 2 - 3 112 73
XM_011537320.2 XP_011535622.1 2 - 3 234 73
XM_011537321.2 XP_011535623.1 2 - 3 198 73
XM_011537322.2 XP_011535624.2 2 - 3 194 73
XM_017021763.1 XP_016877252.1 2 - 3 160 73
XM_017021764.1 XP_016877253.1 2 - 3 173 73
GenBank mRNA AK023979.1 - 2 - 3 100 73
AK055422.1 - 2 - 3 713 73
AK307062.1 - 2 - 3 109 73
BC094003.1 - 2 - 3 269 73
BC094005.1 - 2 - 3 269 73

View Full Product Details